missing translation for 'onlineSavingsMsg'
Learn More

Triethylenetetramine Hydrate, TRC

Product Code. 30451758
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451758 50 g 50g
30452987 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30451758 Supplier Toronto Research Chemicals Supplier No. TRCT77663050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triethylenetetramine Hydrate
Molecular Formula C6 H18 N4 . x(H2 O)
Quantity 50 g
Synonym N1,N2-Bis(2-aminoethyl)-1,2-ethanediamine Hydrate, 1,2-Bis(2-aminoethylamino)ethane Hydrate, 1,4,7,10-Tetraazadecane Hydrate, 1,8-Diamino-3,6-diazaoctane Hydrate, 3,6-Diazaoctane-1,8-diamine Hydrate, Ancamine TETA Hydrate, EpoxySet 145-20020 Hydrate, N,N'-Bis(2-aminoethyl)-1,2-diaminoethane Hydrate, N,N'-Bis(2-aminoethyl)-1,2-ethanediamine Hydrate, N,N'-Bis(2-aminoethyl)ethylenediamine Hydrate, NSC 443 Hydrate, RT 1AX Hydrate, Ren HY 956 Hydrate, Rutapox VE 2896 Hydrate, TECZA Hydrate, TETA Hydrate, Trien Hydrate, Trientine Hydrate, VE 2896, Z1Hydrate
InChI Formula InChI=1S/C6H18N4.H2O/c7-1-3-9-5-6-10-4-2-8;/h9-10H,1-8H2;1H2
SMILES O.NCCNCCNCCN
IUPAC Name N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine;hydrate
Molecular Weight (g/mol) 164.25
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.