Learn More
Triethylene Glycol Dimethyl Ether, Sigma-Aldrich™
Specifications
Specifications
| CAS | 112-49-2 |
| Melting Point | −45°C (Lit.) |
| Density | 0.986 g/mL at 25°C |
| Boiling Point | 216°C |
| Molecular Formula | CH3O(CH2CH2O)3CH3 |
| MDL Number | MFCD00008504 |
| Quantity | 250 mL |
| Synonym | 1,2-Bis(2-methoxyethoxy)ethane, Dimethyltriglycol, NSC 66400, Triglyme, 1,2-Bis(2-methoxyethoxy)ethane, Dimethyltriglycol, NSC 66400, Triglyme |
| Molecular Weight (g/mol) | 178.23 |
| Grade | Research |