missing translation for 'onlineSavingsMsg'
Learn More

Triethylammonium Sulfate, TRC

Product Code. 30456889
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456889 1 g 1g
30455772 100 mg 100mg
30454745 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456889 Supplier Toronto Research Chemicals Supplier No. TRCT7974701G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54272-29-6
Molecular Formula C6H15N .xH2O4S
Molecular Weight (g/mol) 199.27
Synonym N,N-diethylethanamine sulfate
IUPAC Name N,N-diethylethanamine;sulfuric acid
SMILES CCN(CC)CC.OS(=O)(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H15N.H2O4S/c1-4-7(5-2)6-3;1-5(2,3)4/h4-6H2,1-3H3;(H2,1,2,3,4)
Formula Weight 199.0878
Chemical Name or Material Triethylammonium Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.