missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine, Sigma-Aldrich™

Product Code. 15899610
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
40 kg
Unit Size:
1 L
100mL
2.5 L
40kg
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15899610 40 kg 40kg
15886480 100 mL 100mL
15879610 500 mL 500mL
15896480 1 L 1 L
15889610 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15899610 Supplier Sigma Aldrich Supplier No. 8083529040
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

Triethylamine is a volatile organic compound used in several industrial processes such as oxidation, reduction, elimination, substitution, addition, and dehydrohalogenation reactions.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-44-8

Specifications

Melting Point -115°C
Density 0.73 g/cm3 at 20°C
Boiling Point 90°C/1013 hPa
Molecular Formula (C2H5)3N
MDL Number MFCD00009051
Quantity 40 kg
Synonym Triethylamine, N,N-Diethylethanamine, TEA, Triethylamine
Molecular Weight (g/mol) 101.19
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.