Learn More
Triethyl Phosphate-d15, TRC
Specifications
Specifications
| Chemical Name or Material | Triethyl phosphate-d15 |
| CAS | 135942-11-9 |
| Purity Grade Notes | GC |
| Molecular Formula | C6 D15 O4 P |
| Quantity | 50 mg |
| Synonym | Triethyl Phosphate D15, Ethan-d5-ol','phosphate (3:1) (9CI), Triethyl Phosphate D15, Ethan-d5-ol','phosphate (3:1) (9CI) |
| InChI Formula | InChI=1S/C6H15O4P/c1-4-8-11(7,9-5-2)10-6-3/h4-6H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])OP(=O)(OC([2H])([2H])C([2H])([2H])[2H])OC([2H])([2H])C([2H])([2H])[2H] |
| IUPAC Name | tris(1,1,2,2,2-pentadeuterioethyl) phosphate |
| Molecular Weight (g/mol) | 197.25 |
| Show More |