Learn More
Triethyl aconitate, TRC
Product Code. 30454829
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454829 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30454829 Supplier Toronto Research Chemicals Supplier No. TRCT57488050MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Triethyl aconitate
CAS 5349-99-5
Molecular Formula C12H18O6
Quantity 50 mg
Synonym 1-Propene-1,2,3-tricarboxylic acid','triethyl ester, Ethyl aconitate
InChI Formula InChI=1S/C12H18O6/c1-4-16-10(13)7-9(12(15)18-6-3)8-11(14)17-5-2/h7H,4-6,8H2,1-3H3/b9-7+
SMILES C(\CC(OCC)=O)(=C\C(OCC)=O)/C(OCC)=O
IUPAC Name triethyl (E)-prop-1-ene-1,2,3-tricarboxylate
Molecular Weight (g/mol) 258.27
Formula Weight 258.11