missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, Supelco™

Product Code. 32239125
Change view
Click to view available options
Quantity:
25 L
Unit Size:
25 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32239125 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32239125 Supplier Supelco Supplier No. 8223419026
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

EMPLURA™

EMPLURA™ is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triethanolamine
CAS 102-71-6
Melting Point 21°C
Density 1.12 g/cm³ at 20°C
Boiling Point 360°C (1013 hPa) (decomposition)
Flash Point 179°C (closed cup)
Molecular Formula N(CH2CH2OH)3
Quantity 25 L
Synonym Triethanolamine, Tris(2-hydroxyethyl)amine, 2,2′,2′′-Trihydroxytriethylamine, TEA, Triethanolamine
Molecular Weight (g/mol) 149.19
Grade Not Available
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.