missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, Millipore™

Product Code. 10765991
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10765991 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10765991 Supplier Millipore Supplier No. 1083791000
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

GR for analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-71-6

Specifications

Melting Point 21°C
Density 1.12 g/cm3 at 20°C
Boiling Point 360°C/1013 hPa (decomposition)
Molecular Formula N(CH2CH2OH)3
MDL Number MFCD00002855
Quantity 1 L
Synonym Triethanolamine, Tris(2-hydroxyethyl)amine, 2,2′,2′′-Trihydroxytriethylamine, TEA, Triethanolamine
Molecular Weight (g/mol) 149.19
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.