Learn More
Tridemorph, TRC
Product Code. 30134620
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134620 25 g 25g
30132478 10 g 10g
18033510 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134620 Supplier Toronto Research Chemicals Supplier No. TRCT77555025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 24602-86-6
Molecular Formula C19 H39 N O
Molecular Weight (g/mol) 297.52
Synonym Morpholine, 2,6-dimethyl-4-tridecyl-, 2,6-Dimethyl-4-tridecylmorpholine, N-Tridecyl-2,6-dimethylmorpholine, NSC 232676, Tridemorph
IUPAC Name 2,6-dimethyl-4-tridecylmorpholine
SMILES CCCCCCCCCCCCCN1CC(C)OC(C)C1
Specifications
Quantity 25 g
InChI Formula InChI=1S/C19H39NO/c1-4-5-6-7-8-9-10-11-12-13-14-15-20-16-18(2)21-19(3)17-20/h18-19H,4-17H2,1-3H3
Formula Weight 297.3032
Chemical Name or Material N-Tridecyl-2,6-dimethylmorpholine