Learn More
Tricyclohexylchlorotin, TRC
Product Code. 30456829
Change view
Click to view available options
Quantity:
2.5 g
5 g
10 g
Unit Size:
10g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456829 5 g 5g
30454822 10 g 10g
30457233 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30456829 Supplier Toronto Research Chemicals Supplier No. TRCT7745005G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3091-32-5
Molecular Formula C18 H33 Cl Sn
Molecular Weight (g/mol) 403.62
Synonym Tin, chlorotricyclohexyl- (7CI), Tricyclohexyltin chloride (6CI), Chlorotricyclohexylstannane, Chlorotricyclohexyltin, NSC 202692, Tricyclohexylstannyl chloride, Tris(cyclohexyl)tin chloride
IUPAC Name chloro(tricyclohexyl)stannane
SMILES Cl[Sn](C1CCCCC1)(C2CCCCC2)C3CCCCC3
Specifications
Quantity 5 g
InChI Formula InChI=1S/3C6H11.ClH.Sn/c3*1-2-4-6-5-3-1;;/h3*1H,2-6H2;1H;/q;;;;+1/p-1
Formula Weight 404.1293
Chemical Name or Material Tricyclohexyltin chloride