Learn More
Tricyclene, TRC
Product Code. 30455079
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455079 100 mg 100mg
30455078 50 mg 50mg
30450063 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30455079 Supplier Toronto Research Chemicals Supplier No. TRCT797465100MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 508-32-7
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
Synonym 1,7,7-Trimethyltricyclo[2.2.1.02,6]heptane, Cyclene, NSC 86978, Teresantanane
IUPAC Name 6,7,7-trimethyl-2,3,4,5-tetrahydro-1H-tricyclo[2.2.1.0^{2,6}]heptane
SMILES CC1(C)C2CC3C(C2)C13C
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C10H16/c1-9(2)6-4-7-8(5-6)10(7,9)3/h6-8H,4-5H2,1-3H3
Formula Weight 136.1252
Chemical Name or Material Tricyclene