Learn More
Triclopyr, TRC
Product Code. 30011145
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011145 5 g 5g
30003755 10 g 10g
17944495 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011145 Supplier Toronto Research Chemicals Supplier No. TRCT7738205G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 55335-06-3
Molecular Formula C7 H4 Cl3 N O3
Molecular Weight (g/mol) 256.47
Synonym Acetic acid, 2-[(3,5,6-trichloro-2-pyridinyl)oxy]- (ACI), 2-[(3,5,6-Trichloro-2-pyridinyl)oxy]acetic acid (ACI), Acetic acid, [(3,5,6-trichloro-2-pyridinyl)oxy]- (9CI), Acetic acid, [(3,5,6-trichloro-2-pyridyl)oxy]- (6CI, 7CI), 2-[(3,5,6-Trichloropyridinyl)oxy]acetic acid, 3,5,6-TPA, 3,5,6-Trichloro-2-pyridinyloxyacetic acid, 3,5,6-Trichloro-2-pyridinyloxyethanoic acid, 3,5,6-Trichloro-2-pyridyloxyacetic acid, Dowco 233, Garlon, Garlon 2, Garlon 250, Grandstsnd, Maxim, Maxim (auxin), NSC 190671, Redeem, Release, Remedy, Triclopyr
IUPAC Name 2-(3,5,6-trichloropyridin-2-yl)oxyacetic acid
SMILES OC(=O)COc1nc(Cl)c(Cl)cc1Cl
Specifications
Quantity 5 g
InChI Formula InChI=1S/C7H4Cl3NO3/c8-3-1-4(9)7(11-6(3)10)14-2-5(12)13/h1H,2H2,(H,12,13)
Formula Weight 254.9257
Chemical Name or Material Triclopyr