Learn More
Trichloromethyl Chloroformate, Sigma-Aldrich™
Product Code. 15866560
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15866560 100 mL 100mL
15840281 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15866560 Supplier Sigma Aldrich Supplier No. 8181540100
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
for synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 503-38-8
Specifications
Melting Point -57°C
Density 1.63 g/cm3 at 20°C
Boiling Point 128°C/1.013 hPa
MDL Number MFCD00015553
Quantity 100 mL
Synonym Trichloromethyl chloroformate, Diphosgene, Chloroformic acid trichloromethyl ester, Trichloromethyl chloroformate
Molecular Weight (g/mol) 197.83
Grade Research