Learn More
Trichloroacetyl Chloride, Sigma-Aldrich™
Product Code. 15899620
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15899620 250 mL 250mL
15889620 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15899620 Supplier Sigma Aldrich Supplier No. 8083910250
 Restricted Item
Estimated Shipment: 19-11-2026
For synthesis
for synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 76-02-8
Specifications
Melting Point -57°C
Density 1.62 g/cm3 at 20°C
Boiling Point 132°C/1013 hPa (decomposition)
MDL Number MFCD00000792
Quantity 250 mL
Synonym Trichloroacetyl chloride, Trichloroacetic acid chloride, Trichloroacetyl chloride
Molecular Weight (g/mol) 181.83
Grade Research