Learn More
Trichloro Bisphenol S, TRC
Product Code. 30455334
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455334 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455334 Supplier Toronto Research Chemicals Supplier No. TRCT8968665MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Trichloro Bisphenol S
CAS 80396-78-7
Molecular Formula C12H7Cl3O4S
Quantity 5 mg
Synonym 2,2',6-Trichloro-4,4'-sulfonyldiphenol, 2,6-Dichloro-4-((3-chloro-4-hydroxyphenyl)sulfonyl)phenol
InChI Formula InChI=1S/C12H7Cl3O4S/c13-8-3-6(1-2-11(8)16)20(18,19)7-4-9(14)12(17)10(15)5-7/h1-5,16-17H
SMILES Oc1ccc(cc1Cl)S(=O)(=O)c2cc(Cl)c(O)c(Cl)c2
IUPAC Name 2,6-dichloro-4-(3-chloro-4-hydroxyphenyl)sulfonylphenol
Molecular Weight (g/mol) 353.61
Formula Weight 351.9131