missing translation for 'onlineSavingsMsg'
Learn More

Tricaprin, Approx. 99%, MP Biomedicals™

Product Code. 11441882 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441882 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11441882 Supplier MP Biomedicals Supplier No. 0210529705
  • ONLINE EXCLUSIVE PRICE
    293.00 EUR
    278.25 EUR / 5g
    Save 14.75 EUR
    5% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 621-71-6
Assay Percent Range approx.99%
Molecular Formula C33H62O6
Quantity 5 g
Synonym tricaprin, tridecanoin, glycerol tricaprate, glycerol tridecanoate, decanoin, tri, tricapric glyceride, glycerol tricaprin, 1,2,3-tridecanoylglycerol, capric acid triglyceride, glyceryl tricaprate
InChI Key LADGBHLMCUINGV-UHFFFAOYSA-N
SMILES CCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCC)OC(=O)CCCCCCCCC
IUPAC Name 1,3-bis(decanoyloxy)propan-2-yl decanoate
Molecular Weight (g/mol) 554.85
PubChem CID 69310
ChEBI CHEBI:77388
Formula Weight 554.8
Physical Form Solid
Chemical Name or Material Tricaprin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.