Learn More
Tributylamine, Sigma-Aldrich™
Product Code. 15839620
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
40 kg
Unit Size:
1 L
100mL
2.5 L
40kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15839620 40 kg 40kg
15809620 100 mL 100mL
15819620 1 L 1 L
15829620 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15839620 Supplier Sigma Aldrich Supplier No. 8083589040
 Restricted Item
Estimated Shipment: 25-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 102-82-9
Specifications
Melting Point -70°C
Density 0.78 g/cm3 at 20°C
Boiling Point 216°C to 217°C/1013 hPa
Molecular Formula (CH3CH2CH2CH2)3N
MDL Number MFCD00009431
Quantity 40 kg
Synonym Tributylamine, Tributylamine
Molecular Weight (g/mol) 185.35
Grade Research