missing translation for 'onlineSavingsMsg'
Learn More

Tributyl Citrate, Sigma-Aldrich™

Product Code. 15870901
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15870901 1 L 1 L
15860901 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15870901 Supplier Sigma Aldrich Supplier No. 8203501000
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product can replace highly toxic diluents in membrane preparation and thus aligns with Less Hazardous Chemical Syntheses.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-94-1

Specifications

Melting Point -20°C
Density 1.04 g/cm3 at 20°C
Boiling Point 169°C to 171°C/1 hPa
Molecular Formula HOC(COOCH2CH2CH2CH3)(CH2COOCH2CH2CH2CH3)2
MDL Number MFCD00027217
Quantity 1 L
Synonym Tributyl citrate, Citric acid tributyl ester, Tributyl citrate
Molecular Weight (g/mol) 360.44
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.