missing translation for 'onlineSavingsMsg'
Learn More

Treosulfan, MedChemExpress

Product Code. 16489607
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489607 5 mg 5mg
16487106 100 mg 100mg
16463107 50 mg 50mg
16427068 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16489607 Supplier Medchem Express Supplier No. HY16503
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Treosulfan (NSC 39069) is a bifunctional alkylating agent with activity in ovarian cancer and other solid tumor types.

Treosulfan (NSC 39069) is a bifunctional alkylating agent with activity in ovarian cancer and other solid tumor types.

Specifications

Chemical Name or Material Treosulfan
CAS 299-75-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C6H14O8S2
Quantity 5 mg
Synonym NSC 39069 Treosulphan
Solubility Information DMSO : ≥ 100 mg/mL (359.32 mM) ∣H2O : 50 mg/mL (179.66 mM; Need ultrasonic)
SMILES O[C@H]([C@@H](O)COS(C)(=O)=O)COS(C)(=O)=O
Molecular Weight (g/mol) 278.3
Formula Weight 278.3
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.