Learn More
Trenbolone acetate, Lipomed
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Trenbolone acetate
Specifications
Specifications
| Chemical Name or Material | Trenbolone Acetate |
| CAS | 10161-34-9 |
| Analyte or Component Names | Trenbolone Acetate |
| Shipping Condition | Ice Pk. (-20°C) |
| Molecular Formula | C20 H24 O3 |
| Synonym | Estra-4,9,11-trien-3-one, 17β-hydroxy-, acetate (7 CI,8 CI), (17β)-17-(Acetyloxy)estra-4,9,11-trien-3-one, 17β-Acetoxy-Δ4,9,11-estratrien-3-one, 17β-Acetoxyestra-4,9,11-trien-3-one, 17β-Hydroxyestra-4,9,11-trien-3-one 17-acetate, 3-Oxo-17β-acetoxyestra-4,9,11-triene, Component T-H, Component T-S, Finaplix, Finaplix H, Finaplix-S, RU 1697, Trenbolone acetate, Trienbolone acetate |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CC(=O)O[C@H]1 CC[C@H]2 [C@@H]3 CCC4=CC(=O)CCC4=C3C=C[C@]12 C |
| IUPAC Name | [(8 S,13 S,14 S,17 S)-13-methyl-3-oxo-2,6,7,8,14,15,16,17-octahydro-1 H-cyclopenta[a]phenanthren-17-yl] acetate |
| Industry Type | Pharmaceutical |
| Show More |