Learn More
Trelagliptin Succinate, Mikromol™
Description
Trelagliptin Succinate
Specifications
Specifications
| Chemical Name or Material | Trelagliptin Succinate |
| CAS | 1029877-94-8 |
| Analyte or Component Names | Trelagliptin Succinate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18H20FN5O2.C4H6O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Butanedioic acid, compd. with 2-[[6-[(3R)-3-amino-1-piperidinyl]-3,4-dihydro-3-methyl-2,4-dioxo-1(2H)-pyrimidinyl]methyl]-4-fluorobenzonitrile (1:1), SYR 111472 succinate, SYR 472, Trelagliptin succinate |
| InChI Formula | InChI=1S/C18H20FN5O2.C4H6O4/c1-22-17(25)8-16(23-6-2-3-15(21)11-23)24(18(22)26)10-13-7-14(19)5-4-12(13)9-20;5-3(6)1-2-4(7)8/h4-5,7-8,15H,2-3,6,10-11,21H2,1H3;1-2H2,(H,5,6)(H,7,8)/t15-;/m1./s1 |
| SMILES | CN1C(=O)C=C(N2CCC[C@@H](N)C2)N(Cc3cc(F)ccc3C#N)C1=O.OC(=O)CCC(=O)O |
| IUPAC Name | 2-[[6-[(3R)-3-amino-1-piperidyl]-3-methyl-2,4-dioxo-pyrimidin-1-yl]methyl]-4-fluoro-benzonitrile;succinic acid |
| Show More |