Learn More
Trazodone Isobutyl Ether, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(3-chlorophenyl)-4-[3-(2-methylpropoxy)propyl]piperazine |
| CAS | 2470436-00-9 |
| Molecular Formula | C17 H27 Cl N2 O |
| Quantity | 500 mg |
| Synonym | Trazodone Isobutyl Ether Analog |
| InChI Formula | InChI=1S/C17H27ClN2O/c1-15(2)14-21-12-4-7-19-8-10-20(11-9-19)17-6-3-5-16(18)13-17/h3,5-6,13,15H,4,7-12,14H2,1-2H3 |
| SMILES | CC(C)COCCCN1CCN(CC1)c2cccc(Cl)c2 |
| IUPAC Name | 1-(3-chlorophenyl)-4-[3-(2-methylpropoxy)propyl]piperazine |
| Molecular Weight (g/mol) | 310.862 |
| Formula Weight | 310.181 |