Learn More
Thermo Scientific™ Trap Column Cartridges Holders with nanoViper Fittings
Product Code. 13406836 Shop All Thermo Scientific Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Diameter (Metric):
30 μm (fitting)
75 μm (fitting)
missing translation for 'unitSize':
Each
Length (Metric):
5 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) missing translation for 'unitSize' Length (Metric)
13406836 75 μm (fitting) Each -
13264509 30 μm (fitting) Each 5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Product Code. 13406836 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' 164650
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Works with virtually any valve and any column from any manufacturer.
NanoViper connections come preassembled and high pressure tested. Thermo Scientific Trap Column Cartridges Holders with nanoViper fittings do not grab the tubing at a single point. They seal at the tip without using a ferrule. This gives nanoViper fittings their UHPLC, ensuing virtually dead-volume-free capabilities.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 75 μm (fitting)