Learn More
BRAND™ Blaubrand™ Eterna™ AR-GLAS™ Type 3 Class AS Measuring Pipettes
Product Code. p-8000641 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
0.5 mL
1 mL
2 mL
5 mL
10 mL
20 mL
Graduations:
0.01 mL
0.02 mL
0.05 mL
0.1 mL
Tolerance:
±0.006 mL
±0.007 mL
±0.010 mL
±0.03 mL
±0.030 mL
±0.05 mL
±0.070 mL
±0.10 mL
Unit Size:
Pack of 12
Pack of 6
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Graduations Tolerance unitSize
12005312 0.5 mL 0.01 mL ±0.006 mL Pack of 12
11803980 5 mL 0.1 mL ±0.03 mL Pack of 12
11833920 1 mL 0.1 mL ±0.007 mL Pack of 12
12068359 2 mL 0.02 mL ±0.010 mL Pack of 12
11868331 5 mL 0.05 mL ±0.030 mL Pack of 12
11843920 10 mL 0.1 mL ±0.05 mL Pack of 12
11813980 20 mL 0.1 mL ±0.10 mL Pack of 6
10290021 1 mL 0.01 mL ±0.007 mL Pack of 12
10603561 2 mL 0.02 mL ±0.070 mL Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 12005312 Supplier BRAND™ Supplier No. 28405
Only null left
Add to basket
Add to basket
Manufactured from AR-GLAS™ Soda lime glass, this type 3 Class AS Measuring Pipet includes graduations with DE-M marking and is calibrated for total delivery.
  • Manufactured from AR-GLAS™ Soda lime glass.
  • Type 3 (zero on top)
  • DE-M Marking graduations
  • Calibrated to deliver (TD, Ex.)
  • Supplied with batch number and one batch certificate per packing unit
  • USP individual certificate or DAkkS calibration certificate available upon request.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DIN EN ISO 835
Material AR-GLAS
Graduated Yes
Graduations 0.01 mL
Tolerance ±0.006 mL
Volume (Metric) 0.5 mL