Learn More
Deutsch Neumann™ Transparent Polyethylene Tubing
Product Code. 10603491
Change view
Click to view available options
Diameter (Metric) Inner:
2 mm
8 mm
Diameter (Metric) Outer:
2.8 mm
Unit Size:
100m
50m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Diameter (Metric) Outer Unit Size
10603491 2 mm 2.8 mm 100m
10648325 8 mm - 50m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10603491 Supplier Deutsch Neumann™ Supplier No. 3550228
Sample Preparation and Storage Essentials
Use promo code 27996
View all promotions
Estimated Shipment: 30-10-2026
Resistant to acids, bases, alcohols and salt solutions through a temperature of 80°C.
  • Free of smells and flavors
  • Chemically resistant and flexible
  • Provided in lengths of 5m
Specifications
Certifications/Compliance ISO 9001:2008
Diameter (Metric) Inner 2 mm
Material Polyethylene
Diameter (Metric) Outer 2.8 mm
Overall Length (Metric) 100 m
Thickness (Metric) 0.4 mm
Min. Operating Temperature (Metric) 80°C
Packaging Quantity 1 Each
Color Transparent
Max. Operating Temperature (Metric) 80°C