Learn More
Gilac™ Translucent Airtight Box with Blue Lid
Product Code. 17968901
Change view
Click to view available options
Capacity (Metric):
0.4 L
0.8 L
1.2 L
2 L
4 L
8 L
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
17968901 0.4 L Each
17998851 0.8 L Each
17928581 1.2 L Each
17978461 2 L Each
17968971 4 L Each
17918991 8 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17968901 Supplier Gilac™ Supplier No. G604020
  • ONLINE EXCLUSIVE PRICE
    3.72 EUR
    2.61 EUR / Pack of 1
    Save 1.11 EUR
    29% Off
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
Translucent Airtight Box with Blue Lid is suitable for food contact, and its rounded corners facilitate cleaning. The content is visible and recognizable.
  • Translucent airtight box with blue lid suitable for food contact.
  • It is made up of a transparent polypropylene base allowing the contents to be visible and recognizable.
  • Rounded corners to facilitate cleaning and limit retention areas.
  • Blue soft cover, guaranteeing a perfect seal.
  • Light, very resistant and suitable for food contact.
  • Box for preserving food and its flavours while preserving the contents from external odours.
  • Dishwasher safe.
  • Stackable with its lid, discover the complete range made up of 6 formats ranging from 0.4 L to 8 L depending on your needs.
Specifications
Capacity (English) 13.5 oz.
Capacity (Metric) 0.4 L
Closure Color Blue
Closure Type Airtight Lid
Color Translucent
Material Polypropylene
Shape Rectangular
Product Type Airtight Box
Stackable Yes
Dimensions (L x W x H) 128 x 90 x 60 mm
Length (Metric) 128 mm
Length (English) 5 in.
Width (Metric) 90 mm
Width (English) 3.5 in.
Height (Metric) 60 mm
Height (English) 2.4 in.