Learn More
Thermo Scientific™ Transfer Vessels Withdrawal Device
Product Code. 12794716 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
LN2 Transfer Vessels, For Thermo 10
LN2 Transfer Vessels, For Thermo 20
LN2 Transfer Vessels, For Thermo 30
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12794716 LN2 Transfer Vessels, For Thermo 20 Each
10723514 LN2 Transfer Vessels, For Thermo 30 Each
10025753 LN2 Transfer Vessels, For Thermo 10 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12794716 Supplier Thermo Scientific™ Supplier No. AY509X4
  • ONLINE EXCLUSIVE PRICE
    1230.00 EUR
    1160.00 EUR / Pack of 1
    Save 70.00 EUR
    5% Off
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
Easily dispense up to 8L/min. (2.12 gal./min.) without pouring using the Thermo Scientific™ Transfer Vessels Withdrawal Device.
  • Available for 10, 20 and 30L models
  • TRUSTED_SUSTAINABILITY
    Specifications
    Type LN2 Withdrawal Device
    Capacity (English) 1.05 cu. ft.
    Capacity (Metric) 30 L
    For Use With (Equipment) LN2 Transfer Vessels, For Thermo 20