Learn More
Transfer Ribonucleic Acid, MP Biomedicals™
Product Code. 11440871 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kU
500 U
Unit Size:
1ku
500 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440871 1 kU 1ku
11430871 500 U 500 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11440871 Supplier MP Biomedicals Supplier No. 0215653491
  • ONLINE EXCLUSIVE PRICE
    175.00 EUR
    165.90 EUR / 1ku
    Save 9.10 EUR
    5% Off
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Transfer Ribonucleic Acid can be used for a tRNA standard and in reactions involving tRNAs, such as assays for tRNA synthetase activity.
  • One unit will yield an A260 of 1.0 in 1.0 mL water (1 cm light path).
Specifications
Format Vial (powder)
Product Type Transfer Ribonucleic acid
Quantity 1 kU
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.