Learn More
Transfer Ribonucleic Acid, MP Biomedicals™
Product Code. 11440871 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kU
500 U
Unit Size:
1ku
500 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440871 1 kU 1ku
11430871 500 U 500 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11440871 Supplier MP Biomedicals Supplier No. 0215653491
  • ONLINE EXCLUSIVE PRICE
    175.00 EUR
    165.90 EUR / 1ku
    Save 9.10 EUR
    5% Off
 Restricted Item
Estimated Shipment: 08-10-2026
Transfer Ribonucleic Acid can be used for a tRNA standard and in reactions involving tRNAs, such as assays for tRNA synthetase activity.
  • One unit will yield an A260 of 1.0 in 1.0 mL water (1 cm light path).
Specifications
Format Vial (powder)
Product Type Transfer Ribonucleic acid
Quantity 1 kU
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.