missing translation for 'onlineSavingsMsg'
Learn More

Transfer Ribonucleic Acid, MP Biomedicals™

Product Code. 11430871 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kU
500 U
Unit Size:
1ku
500 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430871 500 U 500 units
11440871 1 kU 1ku
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430871 Supplier MP Biomedicals Supplier No. 0215653490

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Transfer Ribonucleic Acid can be used for a tRNA standard and in reactions involving tRNAs, such as assays for tRNA synthetase activity.

  • One unit will yield an A260 of 1.0 in 1.0 mL water (1 cm light path).
TRUSTED_SUSTAINABILITY

Specifications

Format Vial (powder)
Product Type Transfer Ribonucleic Acid
Quantity 500 U

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.