Learn More
Cole-Parmer™ Transfer Pipette
Product Code. 11888641 Shop All Cole Parmer Products
Change view
Click to view available options
Volume (Metric):
6 mL
Unit Size:
Pack of 400
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11888641 6 mL Pack of 400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11888641 Supplier Cole-Parmer™ Supplier No. 0622669
Estimated Shipment: 04-12-2026
Specifications
Material LDPE
For Use With (Application) Life Sciences, Research, Clinical
Graduated No
Quantity 400 Pack
Volume (Metric) 6 mL
Length (Metric) 225 mm
Type Transfer Pipet