Learn More
trans,trans-2,4-Heptadienal (Technical Grade, 90%), TRC
Specifications
Specifications
| CAS | 4313-03-5 |
| Molecular Formula | C7 H10 O |
| Quantity | 1 g |
| Synonym | (2E,4E)-Heptadienal, (2E,4E)-2,4-Heptadienal (ACI), 2,4-Heptadienal, (E,E)- (8CI), (E,E)-2,4-Heptadien-1-al, (E,E)-2,4-Heptadienal, 2-trans-4-trans-Heptadienal, 2E,4E-Heptadienal, all-trans-2,4-Heptadienal, trans,trans-2,4-Heptadienal, trans-2-trans-4-Heptadienal, 2,4-Heptadienal, (2E,4E)- (9CI, ACI) |
| InChI Formula | InChI=1S/C7H10O/c1-2-3-4-5-6-7-8/h3-7H,2H2,1H3/b4-3+,6-5+ |
| SMILES | CC\C=C\C=C\C=O |
| IUPAC Name | (2E,4E)-hepta-2,4-dienal |
| Molecular Weight (g/mol) | 110.15 |
| Formula Weight | 110.0732 |
| Chemical Name or Material | trans,trans-2,4-Heptadienal |