missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (+/-)-trans-N-Boc-4-(4-bromophenyl)pyrrolidine-3-carboxylic acid, 95%

Product Code. 15419535
Change view
Click to view available options
Quantity:
1g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15419535 1g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15419535 Supplier Alfa Aesar™ Supplier No. H52038.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1217829-96-3
Molecular Formula C16H20BrNO4
MDL Number MFCD07363505
Quantity 1g
Synonym 3r,4s-4-4-bromophenyl-1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, trans-1-boc-4-4-bromophenyl-pyrrolidine-3-carboxylic acid, trans-4-4-bromophenyl-1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, trans-4-4-bromophenyl-1-boc-pyrrolidine-3-carboxylic acid, boc-+/--trans-4-4-bromophenyl pyrrolidine-3-carboxylic acid, 3r,4s-4-4-bromophenyl-1-tert, butoxy carbonyl pyrrolidine-3-carboxylic acid, +/--trans-n-boc-4-4-bromophenyl pyrrolidine-3-carboxylic acid, 3r,4s-4-4-bromophenyl-1-tert-butoxycarbonyl-pyrrolidine-3-carboxylic acid
InChI Key MXJJJGPIDKFWKX-OLZOCXBDSA-N
SMILES CC(C)(C)OC(=O)N1CC(C(C1)C(=O)O)C2=CC=C(C=C2)Br
IUPAC Name (3R,4S)-4-(4-bromophenyl)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-3-carboxylic acid
Molecular Weight (g/mol) 370.243
PubChem CID 44828614
Formula Weight 370.24
Percent Purity 95%
Chemical Name or Material (+/-)-trans-N-Boc-4-(4-bromophenyl)pyrrolidine-3-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.