Learn More
BD Difco TranS Bag, Sterile Transport Bags (for 90 mm plates)
Product Code. 15393044
Change view
Click to view available options
Quantity:
100 Pk.
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15393044 100 Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15393044 Supplier BD Difco Supplier No. 257404
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 03-11-2026
Sterile transport bags.
TRUSTED_SUSTAINABILITY
Specifikationer
For Use With (Application) For 90 mm plates
Sterility Sterile
Quantity 100 Pk.
Product Type Specimen Transport Bag
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.