Learn More
trans-5-Chloro-N-[4-(phenylmethoxy)cyclohexyl]pentanamide, TRC
Specifications
Specifications
| Chemical Name or Material | trans-5-Chloro-N-[4-(phenylmethoxy)cyclohexyl]-pentanamide |
| CAS | 98454-45-6 |
| Molecular Formula | C18 H26 Cl N O2 |
| Quantity | 250 mg |
| Synonym | Pentanamide, 5-chloro-N-[4-(phenylmethoxy)cyclohexyl]-, trans- (9CI), trans-5-Chloro-N-[4-(phenylmethoxy)cyclohexyl]pentanamide |
| InChI Formula | InChI=1S/C18H26ClNO2/c19-13-5-4-8-18(21)20-16-9-11-17(12-10-16)22-14-15-6-2-1-3-7-15/h1-3,6-7,16-17H,4-5,8-14H2,(H,20,21)/t16-,17- |
| SMILES | ClCCCCC(=O)N[C@@H]1CC[C@H](CC1)OCc2ccccc2 |
| IUPAC Name | 5-chloro-N-(4-phenylmethoxycyclohexyl)pentanamide |
| Molecular Weight (g/mol) | 323.86 |
| Formula Weight | 323.1652 |