missing translation for 'onlineSavingsMsg'
Learn More

trans-4-n-Propylcyclohexanecarboxylic acid, 99%, Thermo Scientific™

Product Code. 15413998
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15413998 1 g 1g
15423998 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15413998 Supplier Thermo Scientific Alfa Aesar Supplier No. H57536.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38289-27-9
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD00059559
InChI Key QCNUKEGGHOLBES-UHFFFAOYSA-N
Synonym trans-4-propylcyclohexanecarboxylic acid, 4-propylcyclohexanecarboxylic acid, trans-4-n-propylcyclohexanecarboxylic acid, 1s,4r-4-propylcyclohexane-1-carboxylic acid, propylcyclohexanecarboxylicacid, trans-4-propylcyclohexylcarboxylic acid, 4-trans-propyl cyclohexane carboxylic acid, trans-4-n-propyl-cyclohexanecarboxylicacid, trans-4-n-propylcyclohexane carboxylic acid, cyclohexanecarboxylic acid, 4-propyl-, trans
PubChem CID 1490324
IUPAC Name 4-propylcyclohexane-1-carboxylic acid
SMILES CCCC1CCC(CC1)C(=O)O

Specifications

Melting Point 97°C to 100°C
Quantity 1 g
Formula Weight 170.24
Percent Purity 99%
Chemical Name or Material trans-4-n-Propylcyclohexanecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.