Learn More
trans-4-n-Butylcyclohexanecarboxylic acid, 99%, Thermo Scientific™
Product Code. 15433868
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15433868 1 g 1g
15443868 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15433868 Supplier Thermo Scientific Alfa Aesar Supplier No. H57387.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 38289-28-0
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00059560
InChI Key BALGERHMIXFENA-UHFFFAOYSA-N
Synonym trans-4-butylcyclohexanecarboxylic acid, 4-butylcyclohexanecarboxylic acid, trans-4-n-butylcyclohexanecarboxylic acid, 4-butylcyclohexane carboxylic acid, 4-n-butylcyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-butyl, trans-4-butylcyclohexane carboxylic acid, 4-butylcyclohexanecarboxylic acid cis-and trans-mixture, cyclohexanecarboxylic acid, 4-butyl-, trans, enamine_005334
PubChem CID 2060518
IUPAC Name 4-butylcyclohexane-1-carboxylic acid
SMILES CCCCC1CCC(CC1)C(=O)O
Specifications
Quantity 1 g
Formula Weight 184.28
Percent Purity 99%
Chemical Name or Material trans-4-n-Butylcyclohexanecarboxylic acid
compliance-icons
Product Identifier
  • trans-4-n-Butylcyclohexanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: trans-4-n-Butylcyclohexanecarboxylic acid