missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Hydroxy-L-proline Methyl Ester Hydrochloride, TRC

Product Code. 30008295
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008295 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30008295 Supplier Toronto Research Chemicals Supplier No. TRCH952395250MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 40216-83-9
Molecular Formula C6H12ClNO3
Synonym L-Hydroxyproline Methyl Ester Hydrochloride, Methyl (2S,4R)-4-Hydroxy-2-pyrrolidinecarboxylate Hydrochloride, Methyl L-Hydroxyprolinate Hydrochloride, Methyl trans-4-Hydroxy-L-prolinate Hydrochloride, trans-4-Hydroxy-L-proline Methyl Ester Hydrochloride, trans-4-Hydroxyproline Methyl Ester Hydrochloride, 2S,4R)-Methyl 4-hydroxypyrrolidine-2-carboxylate Hydrochloride
InChI Formula InChI=1S/C6H11NO3.ClH/c1-10-6(9)5-2-4(8)3-7-5;/h4-5,7-8H,2-3H2,1H3;1H/t4-,5+;/m1./s1
SMILES Cl.COC(=O)[C@@H]1C[C@@H](O)CN1
IUPAC Name methyl (2S,4R)-4-hydroxypyrrolidine-2-carboxylate;hydrochloride
Molecular Weight (g/mol) 181.62
Quantity 250 mg
Formula Weight 181.0506
Shipping Condition Room Temperature
Chemical Name or Material trans-4-Hydroxy-L-proline Methyl Ester Hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.