missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Hydroxy-L-proline methyl ester hydrochloride, 98%

Product Code. 15469295
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15469295 25 g 25g
15459295 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469295 Supplier Thermo Scientific Alfa Aesar Supplier No. H51726.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 40216-83-9 | C6H12ClNO3 | 181.616 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40216-83-9
Molecular Formula C6H12ClNO3
Molecular Weight (g/mol) 181.616
MDL Number MFCD00080855
InChI Key KLGSHNXEUZOKHH-JBUOLDKXSA-N
Synonym 2s,4r-methyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride, trans-4-hydroxy-l-proline methyl ester hydrochloride, h-hyp-ome.hcl, methyl 2s,4r-4-hydroxypyrrolidine-2-carboxylate hydrochloride, l-4-hydroxyproline methyl ester hydrochloride, hydroxyproline methyl ester hydrochloride, trans-4-hydroxy-l-proline methyl ester hcl, 2s,4r-methyl 4-hydroxypyrrolidine-2-carboxylate hcl, trans-4-hydroxy-l-proline methyl ester, hydrochloride salt, l-proline, 4-hydroxy-, methyl ester, hydrochloride, 4r
PubChem CID 11435319
IUPAC Name methyl (2S,4R)-4-hydroxypyrrolidine-2-carboxylate;hydrochloride
SMILES COC(=O)C1CC(CN1)O.Cl

Specifications

Beilstein 4716932
Quantity 25 g
Formula Weight 181.62
Percent Purity 98%
Chemical Name or Material trans-4-Hydroxy-L-proline methyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.