Learn More
trans-4-((2-Amino-3,5-dibromobenzyl)amino)cyclohexanol hydrochloride, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 23828-92-4 |
| Molecular Formula | C13H19Br2ClN2O |
| MDL Number | MFCD00078932 |
| Quantity | 100 g |
| SMILES | O[C@H]1CC[C@H](NCC2=CC(Br)=CC(Br)=C2N)CC1.Cl |
| Molecular Weight (g/mol) | 414.56 |
| Percent Purity | 97% |
| Chemical Name or Material | trans-4-((2-Amino-3,5-dibromobenzyl)amino)cyclohexanol hydrochloride |