Learn More
trans-3-Phenylallylamine Hydrochloride (Cinnamylamine Hydrochloride), Mikromol™
Product Code. 30051378
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051378 25 mg 25mg
30050768 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30051378 Supplier Mikromol Supplier No. MM1687.010025
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
trans-3-Phenylallylamine Hydrochloride (Cinnamylamine Hydrochloride)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Cinnamylamine Hydrochloride
CAS 5586-89-0
Analyte or Component Names Cinnamylamine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C9 H11 N . Cl H
Certifications/Compliance ISO 17025
Synonym 3-Phenyl-2-Propen-1-amine Hydrochloride (1:1), 2-Propen-1-amine, 3-phenyl-, hydrochloride (9CI), Cinnamylamine, hydrochloride (6CI,7CI,8CI), trans-3-phenylallylamine hydrochloride, Tranylcypromine Related Compound B as Hydrochloride
InChI Formula InChI=1S/C9H11N.ClH/c10-8-4-7-9-5-2-1-3-6-9;/h1-7H,8,10H2;1H/b7-4+;
SMILES Cl.NC\C=C\c1ccccc1
IUPAC Name (E)-3-phenylprop-2-en-1-amine;hydrochloride
Molecular Weight (g/mol) 169.65
Quantity 25 mg
Formula Weight 169.0658 g/mol