missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Aminocyclohexanecarboxylic acid hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. p-7041471
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15457186

Brand: Thermo Scientific Alfa Aesar H62232.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, glycerol. Insoluble in ethanol, chloroform

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 862401-49-8
Molecular Formula C7H14ClNO2
Molecular Weight (g/mol) 179.644
MDL Number MFCD11040475
InChI Key BOUHXQPJYJANEO-KGZKBUQUSA-N
Synonym trans-3-aminocyclohexanecarboxylic acid hydrochloride, 1r,3r-3-aminocyclohexane-1-carboxylic acid hcl, 1r,3r-3-aminocyclohexanecarboxylic acid hydrochloride, 1r,3r-3-aminocyclohexane-1-carboxylic acid hydrochloride, trans-3-aminocyclohexane carboxylic acid hydrochloride, trans-3-aminocyclohexane-1-carboxylic acid hydrochloride
PubChem CID 46172878
IUPAC Name (1R,3R)-3-aminocyclohexane-1-carboxylic acid;hydrochloride
SMILES C1CC(CC(C1)N)C(=O)O.Cl

Specifications

Solubility Information Soluble in water,glycerol. Insoluble in ethanol,chloroform
Quantity 250 mg
Formula Weight 179.64
Percent Purity 97%
Chemical Name or Material trans-3-Aminocyclohexanecarboxylic acid hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.