missing translation for 'onlineSavingsMsg'
Learn More

trans-3-[4-(Boc-amino)cyclohexyl]propionic acid, 97%

Product Code. 15467906
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467906 1 g 1g
15477906 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467906 Supplier Thermo Scientific Alfa Aesar Supplier No. H62844.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 204245-65-8 | C14H25NO4 | 271.357 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 204245-65-8
Molecular Formula C14H25NO4
Molecular Weight (g/mol) 271.357
MDL Number MFCD03844590
InChI Key WCXYTBUDVLMXND-UHFFFAOYSA-N
Synonym cis-3-4-tert-butoxycarbonylaminocyclohexyl propionic acid, cis-3-4-tert-butoxycarbonylaminocyclohexyl-propionic acid, trans-3-4-tert-butoxycarbonylaminocyclohexyl propionic acid, trans-3-4-tert-butoxycarbonylaminocyclohexyl-propionic acid, 3-4-tert-butoxycarbonyl amino cyclohexyl propanoic acid, 3-trans-4-tert-butoxycarbonyl amino cyclohexyl propanoic acid, wcxytbudvlmxnd-phimtyicsa-n, wcxytbudvlmxnd-xypyzodxsa-n, 3-trans-4-tert-butoxycarbonylamino-cyclohexyl-propionic acid
PubChem CID 10516274
IUPAC Name 3-[4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexyl]propanoic acid
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)CCC(=O)O

Specifications

Quantity 1 g
Formula Weight 271.35
Percent Purity 97%
Chemical Name or Material trans-3-[4-(Boc-amino)cyclohexyl]propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.