missing translation for 'onlineSavingsMsg'
Learn More

trans-3-(3-Thienyl)acrylic acid, 98%

Product Code. p-7027942
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485967 5 g 5g
11495967 25 g 25g
11405977 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485967 Supplier Thermo Scientific Alfa Aesar Supplier No. B25017.06
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 102696-71-9 | C7H6O2S | 154.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102696-71-9
Molecular Formula C7H6O2S
Molecular Weight (g/mol) 154.183
MDL Number MFCD00014535
InChI Key VYRYYUKILKRGDN-OWOJBTEDSA-N
Synonym 3-3-thienyl acrylic acid, 3-thien-3-ylacrylic acid, 3-thiophen-3-yl acrylic acid, trans-3-3-thienyl acrylic acid, 2e-3-thiophen-3-yl prop-2-enoic acid, e-3-thiophen-3-yl acrylic acid, 3-thiophen-3-yl prop-2-enoic acid, trans-3-3-thienyl acrylic, 2-propenoic acid, 3-3-thienyl-, 2e, 2e-3-3-thienyl prop-2-enoic acid
PubChem CID 735793
IUPAC Name (E)-3-thiophen-3-ylprop-2-enoic acid
SMILES C1=CSC=C1C=CC(=O)O

Specifications

Melting Point 150°C to 154°C
Quantity 5 g
Beilstein 112245
Formula Weight 154.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material trans-3-(3-Thienyl)acrylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.