missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tramazoline Acetate Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Tramazoline Acetate Salt |
| Molecular Formula | C13H17N3 • C2H4O2 |
| Quantity | 250 mg |
| Synonym | 4,5-Dihydro-N-(5,6,7,8-tetrahydro-1-naphthalenyl)-1H-imidazol-2-amine Acetate Salt, Biciron Acetate Salt, Ellatun Acetate Salt, KB 227 Acetate Salt, Rhinaspray Acetate Salt, Rhinogutt Acetate Salt, Rhinospray Acetate Salt, Rinogutt Acetate Salt, Towk Acetate Salt |
| InChI Formula | InChI=1S/C13H17N3.C2H4O2/c1-2-6-11-10(4-1)5-3-7-12(11)16-13-14-8-9-15-13;1-2(3)4/h3,5,7H,1-2,4,6,8-9H2,(H2,14,15,16);1H3,(H,3,4) |
| SMILES | O=C(C)O.C1(C=CC=C2NC3=NCCN3)=C2CCCC1 |
| IUPAC Name | N-(5,6,7,8-tetrahydronaphthalen-1-yl)-4,5-dihydro-1H-imidazol-2-amine acetate |
| Molecular Weight (g/mol) | 275.34 |
| Formula Weight | 275.16 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?