Learn More
Thermo Scientific™ TraceGOLD™ TG-5MS Guard GC Columns with SafeGuard
Product Code. 11890453 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
15 m
20 m
30 m
Diameter (Metric):
0.18 mm
0.25 mm
0.32 mm
Particle Size:
0.18 μm
0.25 μm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Particle Size Unit Size
11890453 20 m 0.18 mm 0.18 μm Each
11870453 30 m 0.32 mm 0.25 μm Each
11850453 15 m 0.25 mm 0.25 μm Each
11860453 30 m 0.25 mm 0.25 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11890453 Supplier Thermo Scientific™ Supplier No. 260985785
Estimated Shipment: 27-11-2026
Extend column lifetime and get more consistent results over a longer period of time.
Extend column lifetime and get more consistent results over a longer period of time. Thermo Scientific™ TraceGOLD TG-5MS Guard GC Columns with SafeGuard offer the benefit of an integrated SafeGuard to protect the column.
  • With integrated SafeGUARD
  • Available in a range of phases
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Active Compounds Analysis, GC/MS for Analysis of Active Compounds
Particle Size 0.18 μm
Film Thickness 0.18 μm
Length (Metric) 20 m
Polarity Low-Polar
Diameter (Metric) 0.18 mm
USP Type G27, G36
Stationary Phase TraceGOLD™ TG-5 MS
Diameter (Metric) Inner 0.18 mm
Max. Temperature (Metric) 330/350°C (Standard Columns), 430°C (Metal Columns)
Temperature Range (Metric) -60°C to +330/350°C