Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- GC Columns
- GC Capillary Columns
- Thermo Scientific™ TraceGOLD™ TG-5MS GC Columns
Thermo Scientific™ TraceGOLD™ TG-5MS GC Columns
Product Code. p-4522507 Shop All Thermo Scientific Products| Product Code. | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Unit Size |
|---|---|---|---|---|
| 10723297 | 10 m | 0.1 mm | 0.1 μm | Each |
| 15207488 | 12 m | 0.2 mm | 0.33 μm | Each |
| 13408018 | 40 m | 0.15 mm | 0.15 μm | Each |
| 13488008 | 20 m | 0.15 mm | 0.15 μm | Each |
| 15581790 | 30 m | 0.53 mm | 5 μm | Each |
| 10649105 | 15 m | 0.25 mm | 0.25 μm | Each |
| 10066504 | 15 m | 0.32 mm | 0.25 μm | Each |
| 10137334 | 15 m | 0.53 mm | 0.25 μm | Each |
| 10584685 | 30 m | 0.25 mm | 0.25 μm | Each |
| 10086644 | 30 m | 0.32 mm | 0.25 μm | Each |
| 10416954 | 30 m | 0.53 mm | 0.25 μm | Each |
| 10659295 | 60 m | 0.25 mm | 0.25 μm | Each |
| 10289754 | 60 m | 0.32 mm | 0.25 μm | Each |
| 10085884 | 15 m | 0.25 mm | 0.5 μm | Each |
| 10086314 | 15 m | 0.32 mm | 0.5 μm | Each |
| 10096314 | 15 m | 0.53 mm | 0.5 μm | Each |
| 10408874 | 30 m | 0.25 mm | 0.5 μm | Each |
| 10137904 | 30 m | 0.32 mm | 0.5 μm | Each |
| 10457914 | 30 m | 0.53 mm | 0.5 μm | Each |
| 10230005 | 60 m | 0.25 mm | 0.5 μm | Each |
| 10056034 | 60 m | 0.32 mm | 0.5 μm | Each |
| 10584105 | 15 m | 0.32 mm | 1 μm | Each |
| 10594105 | 15 m | 0.53 mm | 1 μm | Each |
| 10650056 | 30 m | 0.25 mm | 1 μm | Each |
| 10681206 | 30 m | 0.32 mm | 1 μm | Each |
| 10744257 | 30 m | 0.53 mm | 1 μm | Each |
| 10576185 | 60 m | 0.25 mm | 1 μm | Each |
| 10586185 | 60 m | 0.32 mm | 1 μm | Each |
| 10753117 | 15 m | 0.53 mm | 1.5 μm | Each |
| 10371025 | 30 m | 0.53 mm | 1.5 μm | Each |
| 12586299 | 60 m | 0.53 mm | 5 μm | Each |
| 11531174 | 20 m | 0.18 mm | 0.18 μm | Each |
Description
Employ the most widely used MS phase in gas chromatography with the 5% phenyl phase TraceGOLD TG-5MS GC Column.
- Low polarity phase, 5% diphenyl/95% dimethyl polysiloxane
- Low bleed for excellent signal-to-noise ratio, sensitivity and mass spectral integrity
- Exceptional inertness ideal for analysis of active compounds
- Equivalent to USP G27 phase
- Similar to Rxi™-5ms, DB-5, HP-5, HP-5ms, Ultra-2, SPB-5, Equity-5, CP-Sil 8
Recommended for:
- Semivolatiles
- Phenols
- Amines
- Residual solvents and solvent impurities
- Drugs of abuse
- Pesticides
- PCB cogeners
- Aroclor mixes
Specifications
Specifications
| For Use With (Application) | Active Compounds Analysis, GC/MS for Analysis of Active Compounds |
| Film Thickness | 0.1 μm |
| Length (Metric) | 10 m |
| Polarity | Low-Polar |
| Description | 0.1 um Thickness; 0.1 mm ID; 10 m Length |
| Diameter (Metric) | 0.1 mm |
| USP Type | G27, G36 |
| Stationary Phase | TraceGOLD™ TG-5 MS |
| Diameter (Metric) Inner | 0.1 mm |
| Product Line | TraceGOLD |
| Max. Temperature (Metric) | 330/350°C |
| Show More Show Less |