Learn More
Thermo Scientific™ TraceGOLD TG-200MS GC Column
Product Code. 15820900 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
30 m
60 m
Diameter (Metric) Inner:
0.25 mm
0.53 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Inner Unit Size
15820900 30 m 0.53 mm Each
15547791 60 m 0.25 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15820900 Supplier Thermo Scientific™ Supplier No. 260842980
Estimated Shipment: 16-11-2026
Thermo Scientific™ TraceGOLD™ TG-200MS GC Column has an exceptionally inert phase with selectivity and elution order optimized for difficult separations.
  • Trifluoropropylmethyl polysiloxane phase resolves compounds that phenyl and cyano phases cannot
  • Outstanding thermal stability and low bleed
  • Suitable for use with sensitive detectors including ECD, NPD and MS
  • Appropriate for use as a confirmation GC column
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 30 m
Diameter (Metric) Inner 0.53 mm
USP Type G6
Stationary Phase TraceGOLD TG-200MS
Film Thickness 1 μm