Learn More
Fisherbrand™ TraceableGO™ Temperature Data Logger with Bluetooth™, Calibration and Bullet Probe
Product Code. 16394446 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Data Memory:
64,000 Readings, 7.5 Months, 5 min. Interval
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Data Memory Unit Size
16394446 64,000 Readings, 7.5 Months, 5 min. Interval Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16394446
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
Easily access data from any location with Bluetooth™ data loggers.
  • Critical Information (Temperature, Trip Duration, MIN/MAX, Battery Level and Alarms) displayed on LCD at all times Simple, single button use
  • Safely transport reagents, clinical samples, vaccines, pharmaceuticals and food items to any location
  • Compatible with TraceableGO or TraceableLIVE Mobile Apps:
    • Once the trip has ended, users may export the data
    • App immediately identifies if the contents experienced conditions outside user-defined parameters
    • View if an alarm was triggered
    • Generate an instant data report
    • Device stores up to 90 alarm events, records readings once every 5 minutes by default, or the user may adjust the reading interval between one minute to once every 12 hours
    • Logging duration is 227 days (7.5 months)
  • TraceableGO™ Bluetooth devices are pre-programmed with alarm values
Specifications
Type Temperature Datalogger
Display Type Multi-line LCD
Data Memory 64,000 Readings, 7.5 Months, 5 min. Interval
Alarm Types High/Low
Software Type TraceableLIVE
Battery Type 2 AAA
Description Bluetooth datalogging thermometer with bullet probe
Includes Batteries, wall mount, Traceable™ Certificate
Channels 1
Certifications/Compliance ISO 17025, NIST
Interface Bluetooth
For Use With (Application) Temperature monitoring
Accuracy (Temperature) ±0.3°C
Temperature Resolution 0.1°
Temperature Range (Metric) -50°C to +70°C
Dimensions (L x W x H) 8.9 x 2 x 5.1 cm
Height (English) 2 in.
Height (Metric) 5.1 cm
Length (English) 8.9 cm
Length (Metric) 3.5 in.
Width (English) 0.8 in.
Width (Metric) 2 cm
Weight (English) 6 oz.
Weight (Metric) 0.17 kg
Show More Show Less
Option to subscribe to cloud-based TraceableLIVE™ service to upload data to cloud.