Learn More
Fisherbrand™ Traceable™ Data Logging Ethernet Thermometers Compatible with TraceableLIVE™ Cloud Service
Product Code. 16526820 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Temperature Range (Metric):
-50°C to +60°C
-90°C to +105°C
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Unit Size
16526820 -50°C to +60°C Each
16536820 -90°C to +105°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16526820
Estimated Shipment: 13-01-2027
Add to basket
Add to basket
Thermometer that allows you to stay connected to your critical environments no matter where you go.
  • Connect to your data via smartphone, tablet, or PC using TraceableLIVE cloud service
  • Get the reliability, speed, and security using Ethernet connection
  • View current temperature conditions anywhere – cloud-based interface sends email, text, and push notifications
  • No software required – web, Android™, Apple™ iOS data visibility
  • Get accurate, reliable measurements with included NIST-traceable calibration
Specifications
Type Thermometer
Calibration Calibrated
Includes One bottle probe with 10 ft. (3 m) long cable, holder with magnetic back, Ethernet cable, universal power adapter and plug with 5 ft. (1.5 m) long cord, three AAA alkaline batteries, and NIST-traceable calibration certificate with data
Material Plastic
For Use With (Application) Temperature monitoring
Temperature Range (Metric) -50°C to +60°C
Length (Metric) 6.985 cm
Weight (Metric) 0.544 kg