Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- GC Columns
- GC Capillary Columns
- Thermo Scientific™ TRACE™ TR-1 GC Columns
Thermo Scientific™ TRACE™ TR-1 GC Columns
Product Code. 10708043 Shop All Thermo Scientific Products| Product Code. | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Unit Size |
|---|---|---|---|---|
| 10708043 | 30 m | 0.32 mm | 1 μm | Each |
| 10668805 | 30 m | 0.53 mm | 1 μm | Each |
| 10279304 | 60 m | 0.32 mm | 0.25 μm | Each |
| 10374363 | 30 m | 0.32 mm | 3 μm | Each |
| 10600174 | 30 m | 0.32 mm | 0.25 μm | Each |
| 10070073 | 30 m | 0.25 mm | 0.5 μm | Each |
| 10311545 | 30 m | 0.53 mm | 5 μm | Each |
| 10269464 | 60 m | 0.32 mm | 1 μm | Each |
| 10496873 | 60 m | 0.25 mm | 1 μm | Each |
| 10585225 | 30 m | 0.32 mm | 0.5 μm | Each |
| 10475533 | 15 m | 0.25 mm | 0.25 μm | Each |
| 10068942 | 15 m | 0.32 mm | 0.25 μm | Each |
| 10475723 | 30 m | 0.25 mm | 0.25 μm | Each |
| 10592635 | 30 m | 0.53 mm | 3 μm | Each |
| 10179414 | 30 m | 0.53 mm | 1.5 μm | Each |
| 10506005 | 100 m | 0.25 mm | 0.5 μm | Each |
| 10725574 | 30 m | 0.53 mm | 0.5 μm | Each |
| 10671206 | 30 m | 0.25 mm | 1 μm | Each |
| 10219264 | 60 m | 0.25 mm | 0.25 μm | Each |
Descrizione
Use this non-polar GC column for method development and analysis of chlorinated and nitroaromatic compounds and environmental samples.
Use the Thermo Scientific™ TRACE™ TR-1 GC column for method development and chlorinated and nitroaromatic compounds and environmental analyses. These non-polar phase, 100% dimethyl polysiloxane gas chromatography columns have a high operating temperature.
Specifica
Specifica
| Film Thickness | 1 μm |
| Length (Metric) | 30 m |
| Stationary Phase | TR-1 |
| Diameter (Metric) Inner | 0.32 mm |
| Product Line | TRACE |